missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate hydrate, Fe 21-23 %, Honeywell Fluka™

Product Code. 15617860 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Poly Bottle
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15617860 500 g Poly Bottle 500g
15607860 250 g Poly Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15617860 Supplier Honeywell-Fluka Supplier No. F1135500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fe 21-23 %

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-10-7 , 7664-93-9
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00064589
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
Synonym Ferric sulfate hydrate
PubChem CID 167265
IUPAC Name iron(3+);trisulfate;hydrate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 98.0000%, 2.0000%
Quantity 500 g
Assay Percent Range 21 to 23%
Linear Formula H2SO4
UN Number UN3260
Formula Weight 399.88
Loss on Drying 7.0 - 20.0%
Packaging Poly Bottle
Assay of Fe
Chemical Name or Material Iron(III) sulfate hydrate, Fe 21-23 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.