missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Sulfate, Technical, Fisher Chemical™

Product Code. 10376040 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10376040 500 g 500g
10050210 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376040 Supplier Fisher Chemical Supplier No. I/1160/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number 149714
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
PubChem CID 167265
IUPAC Name diiron(3+) hydrate trisulfate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Physical Form Solid
Quantity 500 g
Formula Weight 399.88g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Iron(III)sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Sulfuric acid, iron(3+) salt (3:2), hydrate, Sulfuric acid,Ferric sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.