accessibility menu, dialog, popup

UserName

CAS RN 153-78-6

IUPAC Name: 9H-fluoren-2-amine
Synonyms: 2-aminofluorene
Molecular Weight (g/mol): 181.24
Molecular Formula: C13H11N
InChi Key: CFRFHWQYWJMEJN-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(CC3=CC=CC=C23)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results