missing translation for 'onlineSavingsMsg'
Learn More

2-Aminofluorene, 98%

Product Code. 11499797
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499797 25 g 25g
11479797 1 g 1g
11489797 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499797 Supplier Thermo Scientific Alfa Aesar Supplier No. B22769.14
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 153-78-6 | C13H11N | 181.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-78-6
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00001125
InChI Key CFRFHWQYWJMEJN-UHFFFAOYSA-N
Synonym 2-aminofluorene, 2-fluorenamine, aminofluoren, 2-fluoreneamine, fluorene, 2-amino, fluoren-2-amine, 2-fluorenylamine, 2-fluroenylamine, fluoren-2-ylamine, aminofluoren german
PubChem CID 1539
IUPAC Name 9H-fluoren-2-amine
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)N

Specifications

Melting Point 126°C to 130°C
Quantity 25 g
Beilstein 1945861
Formula Weight 181.24
Percent Purity 98%
Chemical Name or Material 2-Aminofluorene
compliance-icons
Product Identifier
  • 2-Aminofluorene
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Fluoren-2-ylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.