accessibility menu, dialog, popup

UserName

CH3OONOPNNNNFPFFFFF(E/Z)

CAS RN 153433-21-7

IUPAC Name: ethyl cyano({[tris(pyrrolidin-1-yl)phosphaniumyl]oxy}imino)formate; hexafluoro-λ⁵-phosphanuide
Synonyms: ethyl cyano({[tris(pyrrolidin-1-yl)phosphaniumyl]oxy}imino)formate hexafluorophosphate
Molecular Weight (g/mol): 527.39
Molecular Formula: C17H29F6N5O3P2
InChi Key: RDWDVLFMPFUBDV-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NO[P+](N1CCCC1)(N1CCCC1)N1CCCC1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)