missing translation for 'onlineSavingsMsg'
Learn More

PyOxim, 97%, Thermo Scientific™

Product Code. 11936691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11936691 5 g 5g
11926691 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936691 Supplier Thermo Scientific Acros Supplier No. 441410050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 153433-21-7
Molecular Formula C17H29F6N5O3P2
Molecular Weight (g/mol) 527.39
MDL Number MFCD18643381
InChI Key RDWDVLFMPFUBDV-UHFFFAOYSA-N
Synonym pyoxim, pyoxime, ethyl cyano hydroxyimino acetato-tri-1-pyrrolidinyl-phosphonium hexafluorophosphate, cyano-ethoxycarbonyl methyleneaminooxy tris 1-pyrrolidinyl phosphonium hexafluorophosphate
PubChem CID 46910530
IUPAC Name [(E)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-tripyrrolidin-1-ylphosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NO[P+](N1CCCC1)(N1CCCC1)N1CCCC1)C#N

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 527.39
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material PyOxim
Product Identifier
  • PyOxim

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.