missing translation for 'onlineSavingsMsg'
Learn More

PyOxim, TRC

Product Code. 30419847
Change view
Click to view available options
Quantity:
100 mg
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419847 100 mg
30419068 2.5 g
30413126 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419847 Supplier Toronto Research Chemicals Supplier No. TRCP999410100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153433-21-7
Molecular Formula C17H29F6N5O3P2
Molecular Weight (g/mol) 527.4
Synonym Pyoxime, (T-4)-[ethyl cyano(hydroxyimino)acetato-O2]tri-1-pyrrolidinyl-Phosphorus(1+) hexafluorophosphate(1-), (T-4)-[ethyl 2-cyano-2-[(hydroxy-kO)imino]acetato]tri-1-pyrrolidinyl-Phosphorus(1+) hexafluorophosphate(1-)
IUPAC Name [(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-tripyrrolidin-1-ylphosphanium;hexafluorophosphate
SMILES CCOC(=O)C(=NO[P+](N1CCCC1)(N2CCCC2)N3CCCC3)C#N.F[P-](F)(F)(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H29N5O3P.F6P/c1-2-24-17(23)16(15-18)19-25-26(20-9-3-4-10-20,21-11-5-6-12-21)22-13-7-8-14-22;1-7(2,3,4,5)6/h2-14H2,1H3;/q+1;-1
Formula Weight 527.165
Chemical Name or Material PyOxim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.