CAS RN 155836-50-3
CAS RN 155836-50-3
IUPAC Name:
4-{[2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](3-hydroxyphenyl)methyl}-N,N-diethylbenzamide
Synonyms:
4-{[2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](3-hydroxyphenyl)methyl}-N,N-diethylbenzamide
Molecular Weight (g/mol):
435.61
Molecular Formula:
C27H37N3O2
InChi Key:
LBLDMHBSVIVJPM-UHFFFAOYNA-N
SMILES:
CCN(CC)C(=O)C1=CC=C(C=C1)C(N1CC(C)N(CC=C)CC1C)C1=CC(O)=CC=C1