missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BW 373U86, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Potent, selective non-peptide δ-opioid receptor agonist. Ki values are 1.8, 15 and 34 nM for δ, μ and κ receptors respectively. Centrally active following systemic administration in vivo.
Specifications
Specifications
| CAS | 155836-50-3 |
| Molecular Formula | C27H37N3O2 |
| Quantity | 10 mg |
| Synonym | snc-86, chembl25230, 4-r-2s,5r-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl-3-hydroxyphenyl methyl-n,n-diethylbenzamide, tocris-1663, d0k5kl, +--4-alpha-r*-alpha-2s*,5r*-4-allyl-2,5-dimethyl-1-piperazinyl-3-hydroxybenzyl-n,n-diethylbenzamide, 4-4-allyl-2,5-dimethyl-piperazin-1-yl-3-hydroxy-phenyl-methyl-n,n-diethyl-benzamide, benzamide, 4-2,5-dimethyl-4-2-propenyl-1-piperazinyl 3-hydroxyphenyl methyl-n,n-diethyl-, 1 s* ,2alpha,5beta-+-, +-4-r-2s,5r-2,5-dimethyl-4-2-propenyl-1-piperazinyl-3-hydroxyphenyl methyl-n,n-diethylbenzamide, +/--4-alphar*-alpha 2s*,5r*-4-allyi-2,5-dimethyl-1-piperazinyl-3-hydroxybenzyl-n,n-diethylbenzamide |
| InChI Key | LBLDMHBSVIVJPM-YZIHRLCOSA-N |
| SMILES | CCN(CC)C(=O)C1=CC=C(C=C1)C(C2=CC(=CC=C2)O)N3CC(N(CC3C)CC=C)C |
| IUPAC Name | 4-[(R)-[(2S,5R)-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl]-(3-hydroxyphenyl)methyl]-N,N-diethylbenzamide |
| Molecular Weight (g/mol) | 435.612 |
| PubChem CID | 119029 |
| Formula Weight | 435.61 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction