CAS RN 15727-65-8
CAS RN 15727-65-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Ethynylnaphthalene, MedChemExpress
MedChemExpress 1-Ethynylnaphthalene is a selective inhibitor of cytochrome P450 1B1.