accessibility menu, dialog, popup

UserName

CAS RN 157652-31-8

CH3FFOOH

CAS RN 157652-31-8

IUPAC Name: 3,4-difluoro-2-methylbenzoic acid
Synonyms: 3,4-difluoro-2-methylbenzoic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: AGUUCAUQWFAFPR-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(F)=C1F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results