missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-2-methylbenzoic acid, 97+%, Thermo Scientific™

Product Code. 15495037
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15495037

Brand: Thermo Scientific Alfa Aesar H32326.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,4-Difluoro-2-methylbenzoic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in medicine field, electronic industry and polymer material.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157652-31-8
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD07777153
InChI Key AGUUCAUQWFAFPR-UHFFFAOYSA-N
PubChem CID 12137678
IUPAC Name 3,4-difluoro-2-methylbenzoic acid
SMILES CC1=C(C=CC(F)=C1F)C(O)=O

Specifications

Melting Point 152°C to 156°C
Color White to Cream
Odor Odorless
Quantity 1 g
Synonym 3,4-difluoro-2-methylbenzoicacid, 3,4-difluoro-2-methyl-benzoic acid, 3,4-difluoro-o-toluic acid, benzoic acid, 3,4-difluoro-2-methyl, pubchem8240, benzoic acid,3,4-difluoro-2-methyl
Solubility Information Insoluble in water.
Formula Weight 172.13
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 3,4-Difluoro-2-methylbenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.