accessibility menu, dialog, popup

UserName

CH3OOIOH

CAS RN 157942-12-6

IUPAC Name: methyl 3-hydroxy-4-iodobenzoate
Synonyms: methyl 3-hydroxy-4-iodobenzoate
Molecular Weight (g/mol): 278.05
Molecular Formula: C8H7IO3
InChi Key: LXCQVWRESZDFGW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=C(I)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results