missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxy-4-iodobenzoate, 97%, Thermo Scientific™

Product Code. 15451519
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451519 250 mg 250mg
15431519 1 g 1g
15441519 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451519 Supplier Thermo Scientific Alfa Aesar Supplier No. H64354.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157942-12-6
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD06203960
InChI Key LXCQVWRESZDFGW-UHFFFAOYSA-N
Synonym methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate
PubChem CID 10378812
IUPAC Name methyl 3-hydroxy-4-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)O

Specifications

Quantity 250 mg
Beilstein 7289690
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 278.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-hydroxy-4-iodobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.