CAS RN 1621175-65-2
CAS RN 1621175-65-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ TC-G 1008
Potent and selective GPR39 agonist
2
TC-G-1008, MedChemExpress
MedChemExpress TC-G-1008 (GPR39-C3) is a potent and orally available GPR39 agonist with EC50 values of 0.4 and 0.8 nM for rat and human receptors respectively.
3
4