Learn More
Tocris Bioscience™ TC-G 1008
Potent and selective GPR39 agonist
Brand: Tocris Bioscience™ 5355/10
Description
Potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Reduces TNF-α-induced oxidative stress, mitochondrial dysfunction, cytokine expression and secretion of MMPs in fibroblast-like synoviocytes (FLS). Orally bioavailable.
TC-G 1008 is a potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Reduces TNF-α-induced oxidative stress, mitochondrial dysfunction, cytokine expression and secretion of MMPs in fibroblast-like synoviocytes (FLS). Orally bioavailable.Specifications
| TC-G 1008 | |
| N-[3-Chloro-4-[[[2-(methylamino)-6-(2-pyridinyl)-4-pyrimidinyl]amino]methyl]phenyl]methanesulfonate | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 1621175-65-2 | |
| 10 mg | |
| Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research Use Only. Not For Human or Veterinary Use