CAS RN 1630936-95-6
CAS RN 1630936-95-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
6
of
6
results
1
Tocris Bioscience™ VU 0422288
Selective group III mGlu positive allosteric modulator
2
VU 0422288, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
VU 0422288, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
VU 0422288, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
6