Learn More
Tocris Bioscience™ VU 0422288
GREENER_CHOICE
Selective group III mGlu positive allosteric modulator
Brand:  Tocris Bioscience™ 5378/10
Encompass
Product Code. 15796766
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
VU 0422288 is a group III mGlu receptor positive allosteric modulator (EC50 values are 108, 125 and 146 nM for mGlu4, mGlu8 and mGlu7 respectively). Exhibits selectivity over a panel of 68 other GPCRs, ion channels and transporters. Potentiates mGlu7-mediated reductions in excitatory post synaptic potentials in hippocampal slices.
Specifications
Specifications
VU 0422288
N-[3-Chloro-4-[(5-chloro-2-pyridinyl)oxy]phenyl]-2-pyridinecarboxamide
Soluble to 100mM in DMSO
Bioactive Small Molecule
1630936-95-6
10 mg
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Title
Select an issue
Research Use Only. Not For Human or Veterinary Use