CAS RN 1641-17-4
CAS RN 1641-17-4
IUPAC Name:
5-methoxy-2-(4-methylbenzoyl)phenol
Synonyms:
mexenone benzophenone-10
Molecular Weight (g/mol):
242.27
Molecular Formula:
C15H14O3
InChi Key:
MJVGBKJNTFCUJM-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C(=O)C2=CC=C(C)C=C2)C(O)=C1
1
–
1
of
1
results
1
Mexenone, MedChemExpress
MedChemExpress Mexenone is a potent benzophenone-type UV filter. Mexenone is used for sunscreening agent.