accessibility menu, dialog, popup

UserName

FFFNCS

CAS RN 1645-65-4

IUPAC Name: 1-isothiocyanato-4-(trifluoromethyl)benzene
Synonyms: 1-isothiocyanato-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 203.18
Molecular Formula: C8H4F3NS
InChi Key: DQEVDFQAYLIBRD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results