missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenyl isothiocyanate, 98+%

Product Code. 11342697
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342697 5 g 5g
11332697 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342697 Supplier Thermo Scientific Alfa Aesar Supplier No. L11890.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1645-65-4 | C8H4F3NS | 203.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1645-65-4
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
MDL Number MFCD00039645
InChI Key DQEVDFQAYLIBRD-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethyl benzene, 4-trifluoromethyl phenylisothiocyanate, 4-trifluoromethylphenylisothiocyanate, p-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolyl isothiocyanate, isothiocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethyl-benzene
PubChem CID 137134
IUPAC Name 1-isothiocyanato-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)N=C=S

Specifications

Melting Point 39°C to 42°C
Boiling Point 207°C
Flash Point >110°C (230°F)
Odor Pungent
Quantity 5 g
UN Number UN2923
Beilstein 2693553
Sensitivity Moisture sensitive
Formula Weight 203.19
Percent Purity ≥98%
Chemical Name or Material 4-(Trifluoromethyl)phenyl isothiocyanate
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)phenyl isothiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-(Trifluoromethyl)phenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.