accessibility menu, dialog, popup

UserName

CH3OH2NNOO

CAS RN 16554-45-3

IUPAC Name: 2-methoxy-6-nitroaniline
Synonyms: 2-methoxy-6-nitroaniline
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: NDKWDGCTUOOAPF-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1N)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results