missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-nitroaniline, 97%, Thermo Scientific™

Product Code. 15457577
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457577 1 g 1g
15467577 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457577 Supplier Thermo Scientific Alfa Aesar Supplier No. H35777.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16554-45-3
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD01930197
InChI Key NDKWDGCTUOOAPF-UHFFFAOYSA-N
Synonym 2-methoxy-6-nitrophenylamine, 2-amino-3-nitroanisole, 2-methoxy-6-nitro-phenylamine, 6-nitro-o-anisidine, 2-methoxy-6-nitrobenzenamine, 6-methoxy-2-nitroaniline, 2-amino-3-methoxynitrobenzene, benzenamine, 2-methoxy-6-nitro, ambkkkkk250, pubchem19661
PubChem CID 85491
IUPAC Name 2-methoxy-6-nitroaniline
SMILES COC1=CC=CC(=C1N)[N+](=O)[O-]

Specifications

Melting Point 73°C to 78°C
Quantity 1 g
UN Number UN2811
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-6-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.