accessibility menu, dialog, popup

UserName

OHPOHOCl

CAS RN 16672-87-0

IUPAC Name: (2-chloroethyl)phosphonic acid
Synonyms: ethephon 2-chloraethyl-phosphonsaeure 2-cepa
Molecular Weight (g/mol): 144.49
Molecular Formula: C2H6ClO3P
InChi Key: UDPGUMQDCGORJQ-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCCl
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)