missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl) Phosphonic Acid (>85%), TRC

Product Code. 30138369
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138369 100 mg 100mg
30133426 25 g 25g
18012940 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138369 Supplier Toronto Research Chemicals Supplier No. TRCC366175100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16672-87-0
Molecular Formula C2 H6 Cl O3 P
Molecular Weight (g/mol) 144.49
IUPAC Name 2-chloroethylphosphonic acid
SMILES OP(=O)(O)CCCl

Specifications

Quantity 100 mg
Synonym Ethephon, Phosphonic acid, P-(2-chloroethyl)- (ACI), P-(2-Chloroethyl)phosphonic acid (ACI), Phosphonic acid, (2-chloroethyl)- (7CI, 8CI, 9CI), (2-Chloroethyl)phosphonic acid, 2-CEPA, 2-Chloroethanephosphonic acid, 2-KhEFK, Amchem 68-250, Arvest, Boll'd, Camposan, Camposan M, CEP, CEPA, CEPA (pesticide), Cepha 10LS, Cerone, Chlorethephon, Chloroethylphosphonic acid, Dextrel M, Direphon, Ethefon, Ethel, Ethephon, Ethereal, Etheverse, Ethrel, Ethrel 10, Ethrel 48 SL, Ethrel 48SL, Exin, Flordimex, Flordimex T, Flordimex TM, Florel, Fruitel, G 996, G 996 (pesticide), Hevetex, Kamposan, KhEFK, MATURE-AIDE, Mature-Aide, MFX 0307, MFX 0543, Prep, Prep (growth regulator), PRM 12, PRM 12RP, Proxy, Roll-Fruct, Romtrel, Super Boll, Tomathrel, β-Chloroethylphosphonic acid
InChI Formula InChI=1S/C2H6ClO3P/c3-1-2-7(4,5)6/h1-2H2,(H2,4,5,6)
Formula Weight 143.9743
Chemical Name or Material Ethephon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.