accessibility menu, dialog, popup

UserName

NBrOO

CAS RN 166953-64-6

IUPAC Name: benzyl 4-bromopiperidine-1-carboxylate
Synonyms: benzyl 4-bromopiperidine-1-carboxylate
Molecular Weight (g/mol): 298.18
Molecular Formula: C13H16BrNO2
InChi Key: BFWKSPOSMPKTSV-UHFFFAOYSA-N
SMILES: BrC1CCN(CC1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results