missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-bromotetrahydro-1(2H)-pyridinecarboxylate, ≥97%, Thermo Scientific™

Product Code. 10460734
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10460734 5 g 5g
10334092 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10460734 Supplier Thermo Scientific Maybridge Supplier No. CC07510DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 166953-64-6
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD02681990
InChI Key BFWKSPOSMPKTSV-UHFFFAOYSA-N
Synonym benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
PubChem CID 2776275
IUPAC Name benzyl 4-bromopiperidine-1-carboxylate
SMILES C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.374g/mL
Assay Percent Range 97%
Quantity 5 g
Formula Weight 298.18
Percent Purity 97%
Chemical Name or Material Benzyl 4-bromotetrahydro-1(2H)-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.