missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-bromopiperidine-1-carboxylate, 96%

Product Code. 10193444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10193444 5 g Glass bottle 5g
10133404 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193444 Supplier Thermo Scientific Acros Supplier No. 427030050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 166953-64-6 | C13H16BrNO2 | 298.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 166953-64-6
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD02681990
InChI Key BFWKSPOSMPKTSV-UHFFFAOYSA-N
Synonym benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
PubChem CID 2776275
IUPAC Name benzyl 4-bromopiperidine-1-carboxylate
SMILES C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2

Spécification

CAS Min % 95.0
CAS Max % 100.0
Density 1.3740g/mL
Boiling Point 144°C (0.3 mmHg)
Flash Point >110°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.374
Formula Weight 298.18
Percent Purity 96%
Chemical Name or Material Benzyl 4-bromopiperidine-1-carboxylate
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Benzyl 4-bromopiperidine-1-carboxylate
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.