accessibility menu, dialog, popup

UserName

H3COClHO

CAS RN 16766-30-6

IUPAC Name: 4-chloro-2-methoxyphenol
Synonyms: 4-chloroguaiacol
Molecular Weight (g/mol): 158.58
Molecular Formula: C7H7ClO2
InChi Key: FVZQMMMRFNURSH-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=CC=C1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)