missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxyphenol, 97%

Product Code. 11442557
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442557 25 g 25g
11432557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442557 Supplier Thermo Scientific Alfa Aesar Supplier No. A14806.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16766-30-6 | C7H7ClO2 | 158.58 g/mol

4-Chloro-2-methoxyphenol, is used as an intermediate in pharmaceutical research, pharmaceutical chemicals and reagents. It is also used as an application intermediate. it can be used to produce 4-Chloro-2-methoxy-6-nitrophenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-2-methoxyphenol, is used as an intermediate in pharmaceutical research, pharmaceutical chemicals and reagents. It is also used as an application intermediate. it can be used to produce 4-Chloro-2-methoxy-6-nitrophenol.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16766-30-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.58
MDL Number MFCD00070774
InChI Key FVZQMMMRFNURSH-UHFFFAOYSA-N
PubChem CID 28050
IUPAC Name 4-chloro-2-methoxyphenol
SMILES COC1=CC(Cl)=CC=C1O

Specifications

Melting Point 16°C to 17°C
Density 1.304
Boiling Point 241°C
Flash Point >110°C (230°F)
Odor Phenol-like
Refractive Index 1.5595
Quantity 25 g
Beilstein 1940516
Synonym 4-chloroguaiacol, phenol, 4-chloro-2-methoxy, 4-chloro-2-methoxy-phenol, 2-methoxy-4-chloro-phenol, pubchem3665, acmc-209dwx, #, phenol,4-chloro-2-methoxy, 4-chloranyl-2-methoxy-phenol, fvzqmmmrfnursh-uhfffaoysa
Solubility Information It is slightly soluble in water.
Formula Weight 158.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.