accessibility menu, dialog, popup

UserName

H3CSBOHHO

CAS RN 168618-42-6

IUPAC Name: [2-(methylsulfanyl)phenyl]boronic acid
Synonyms: 2-(methylsulfanyl)phenylboronic acid
Molecular Weight (g/mol): 168.02
Molecular Formula: C7H9BO2S
InChi Key: QXBWTYBCNFKURT-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results