missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzeneboronic acid, 98+%

Product Code. 11398426
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398426 1 g 1g
11308436 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398426 Supplier Thermo Scientific Alfa Aesar Supplier No. L17456.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 168618-42-6 | C7H9BO2S | 168.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168618-42-6
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.02
MDL Number MFCD01318165
InChI Key QXBWTYBCNFKURT-UHFFFAOYSA-N
Synonym 2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid, 2-methylsulfanylphenyl boronic acid, 2-methylsulfanyl phenylboronic acid, 2-methylsulfanyl phenyl boranediol, 2-methylsulfanyl phenyl boronic acid, o-methylthio phenylboronic acid
PubChem CID 2773543
IUPAC Name (2-methylsulfanylphenyl)boronic acid
SMILES CSC1=CC=CC=C1B(O)O

Specifications

Melting Point ∼160°C
Quantity 1 g
Beilstein 8309529
Formula Weight 168.02
Percent Purity ≥98%
Chemical Name or Material 2-(Methylthio)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.