CAS RN 16962-40-6
CAS RN 16962-40-6
IUPAC Name:
titanium(4+) diammonium hexafluoride
Synonyms:
titanium(4+) diammonium hexafluoride
Molecular Weight (g/mol):
197.94
Molecular Formula:
F6H8N2Ti
InChi Key:
NMGYKLMMQCTUGI-UHFFFAOYSA-J
SMILES:
[NH4+].[NH4+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
1
–
8
of
8
results
1
Ammonium hexafluorotitanate, 99.99% (metals basis)
CAS: 16962-40-6 Molecular Formula: (NH4)2TiF6 MDL Number: MFCD00010888
2
Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis)
CAS: 16962-40-6 Molecular Formula: H8F6N2Ti Molecular Weight (g/mol): 197.97 MDL Number: MFCD00010888
3
8