missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorotitanate, 99.99% (metals basis)

Product Code. 11324898
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324898 500 g 500g
11304898 25 g 25g
11314898 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324898 Supplier Thermo Scientific Alfa Aesar Supplier No. 043192.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16962-40-6 | (NH4)2TiF6

Ammonium hexafluorotitanate is used as an anti-corrosive cleaning agent. It is also used for the production of ceramics and glass. Further, it is used in the preparation of synthetic gems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16962-40-6
Molecular Formula (NH4)2TiF6
MDL Number MFCD00010888

Specifications

Color White
Physical Form Crystalline
Quantity 500 g
Solubility Information Insoluble in water.
Formula Weight 197.95
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Ammonium hexafluorotitanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.