accessibility menu, dialog, popup

UserName

CAS RN 16965-06-3

CH3H3CCH3OONH(S)NHOFFFHOO

CAS RN 16965-06-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-6-(2,2,2-trifluoroacetamido)hexanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-6-(2,2,2-trifluoroacetamido)hexanoic acid
Molecular Weight (g/mol): 342.32
Molecular Formula: C13H21F3N2O5
InChi Key: DEIYNDIFGSDDCY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCCNC(=O)C(F)(F)F)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)