accessibility menu, dialog, popup

UserName

CAS RN 17095-24-8

IUPAC Name: tetrasodium (3E)-5-amino-4-oxo-6-(2-{4-[2-(sulfonatooxy)ethanesulfonyl]phenyl}diazen-1-yl)-3-(2-{4-[2-(sulfonatooxy)ethanesulfonyl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: tetrasodium (6E)-4-amino-5-oxo-3-(2-{4-[2-(sulfonatooxy)ethanesulfonyl]phenyl}diazen-1-yl)-6-(2-{4-[2-(sulfonatooxy)ethanesulfonyl]phenyl}hydrazin-1-ylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 991.79
Molecular Formula: C26H21N5Na4O19S6
InChi Key: FWKNDLPEBIXZRV-FXJKTVKKSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].NC1=C(N=NC2=CC=C(C=C2)S(=O)(=O)CCOS([O-])(=O)=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)S(=O)(=O)CCOS([O-])(=O)=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results