missing translation for 'onlineSavingsMsg'
Learn More

C.I.Reactive Black 5 (Technical Grade), TRC

Product Code. 30136408
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136408 5 g 5g
30010445 500 mg 500mg
30008159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136408 Supplier Toronto Research Chemicals Supplier No. TRCC2251855G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17095-24-8
Molecular Formula C26 H21 N5 O19 S6 . 4 Na
Molecular Weight (g/mol) 991.82
Synonym 2,7-Naphthalenedisulfonic acid, 4-amino-5-hydroxy-3,6-bis[2-[4-[[2-(sulfooxy)ethyl]sulfonyl]phenyl]diazenyl]-, sodium salt (1:4) (ACI), 2,7-Naphthalenedisulfonic acid, 4-amino-5-hydroxy-3,6-bis[[4-[[2-(sulfooxy)ethyl]sulfonyl]phenyl]azo]-, tetrasodium salt (9CI), 2,7-Naphthalenedisulfonic acid, 4-amino-5-hydroxy-3,6-bis[[p-[(2-hydroxyethyl)sulfonyl]phenyl]azo]-, 3,6-bis(hydrogen sulfate) (ester), tetrasodium salt (8CI), Active black RB 5, Adizol Black B, Atulzol Black B, Basilen Black F-B, Basilen Black F-DF, C.I. 20505, C.I. Reactive Black 5, Cavalite Black B, Celmazol Black B, Cibacron Black WNN 2R, Cibacron Marine W-B, Cibacron Navy DP-B, Cibacron Navy W-B, Diamira Black B, Drimarene Black R/K 3B, Duractive Black B, Everzol Black B, Everzol Black BHC, Everzol Black GSP, Helaktyn Black B, Intracron Black VS-B, Ismative Black B, Jackazol Black B, Levafix Black E-B, Navacron Black R, Navictive Black B, Novacron Black R, Novacron Navy W-B, Novacron Navy W-B IN, Ostazin Black V-B, Primazin Black BN, Primazine Black BN, Procion Black E-B, RBk 5, Reactive Black 4ST, Reactive Black 5, Reactive Black B, Reactive Black B 5, Reactive Black B Plus, Reactive Black HBL, Reactive Black KN-G 2RC, Reactive Black KN-GRRC, Reactive Black KNB, Reactive Black V 2B, Reactive Black VB, Reactive Dark Blue B-BF, Remazol Black 133B, Remazol Black 5, Remazol Black B, Remazol Black B 133, Remazol Black B-ID, Remazol Black GF, Remazol Black GR, Remazol Black SB, Ruikesu Black B 133, Setazol Black DPT, Sumifix Black B, Sumifix Supra Black B, Synozol Black B, Synozol Black B 150, Taifix Black B-T
IUPAC Name tetrasodium;4-amino-5-hydroxy-3,6-bis[(E)-[4-(2-sulfonatooxyethylsulfonyl)phenyl]diazenyl]naphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].Nc1c(N=Nc2ccc(cc2)S(=O)(=O)CCOS(=O)(=O)[O-])c(cc3cc(c(N=Nc4ccc(cc4)S(=O)(=O)CCOS(=O)(=O)[O-])c(O)c13)S(=O)(=O)[O-])S(=O)(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C26H25N5O19S6.4Na/c27-23-22-15(13-20(53(37,38)39)24(23)30-28-16-1-5-18(6-2-16)51(33,34)11-9-49-55(43,44)45)14-21(54(40,41)42)25(26(22)32)31-29-17-3-7-19(8-4-17)52(35,36)12-10-50-56(46,47)48;;;;/h1-8,13-14,32H,9-12,27H2,(H,37,38,39)(H,40,41,42)(H,43,44,45)(H,46,47,48);;;;/q;4*+1/p-4/b30-28+,31-29+;;;;
Formula Weight 990.8746
Chemical Name or Material Reactive Black 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.