accessibility menu, dialog, popup

UserName

OHH2C

CAS RN 1745-81-9

IUPAC Name: 2-(prop-2-en-1-yl)phenol
Synonyms: 2-allylphenol
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: QIRNGVVZBINFMX-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1CC=C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results