missing translation for 'onlineSavingsMsg'
Learn More

2-Allylphenol, 98+%

Product Code. 11449642
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449642 500 g 500g
11439642 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449642 Supplier Thermo Scientific Alfa Aesar Supplier No. A10755.36
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1745-81-9 | C9H10O | 134.178 g/mol

2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

Solubility
Soluble in water(7 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1745-81-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002250
InChI Key QIRNGVVZBINFMX-UHFFFAOYSA-N
Synonym 2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol
PubChem CID 15624
ChEBI CHEBI:39826
IUPAC Name 2-prop-2-enylphenol
SMILES C=CCC1=CC=CC=C1O

Specifications

Melting Point -6°C
Density 1.024
Boiling Point 220°C to 222°C
Flash Point 88°C (190°F)
Odor Phenol-like
Refractive Index 1.545
Quantity 500 g
UN Number UN2922
Beilstein 742121
Solubility Information Soluble in water(7g/L).
Formula Weight 134.18
Percent Purity ≥98%
Chemical Name or Material 2-Allylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.