accessibility menu, dialog, popup

UserName

CAS RN 175136-78-4

IUPAC Name: 2,4-dichloro-1-(2-iodophenoxy)benzene
Synonyms: 2,4-dichloro-1-(2-iodophenoxy)benzene
Molecular Weight (g/mol): 364.99
Molecular Formula: C12H7Cl2IO
InChi Key: ULAWXTPGHWKSDY-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(OC2=CC=CC=C2I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results