missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-(2-iodophenoxy)benzene, ≥95%, Thermo Scientific™

Product Code. 10762863
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762863 1 g Amber Glass Bottle 1g
10621103 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10762863 Supplier Thermo Scientific Maybridge Supplier No. CD08469DA
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175136-78-4
Molecular Formula C12H7Cl2IO
Molecular Weight (g/mol) 364.99
MDL Number MFCD00052444
InChI Key ULAWXTPGHWKSDY-UHFFFAOYSA-N
Synonym 2,4-dichloro-1-2-iodophenoxy benzene, 2,4-dichloro-1 2-iodophenoxy benzene, benzene,2,4-dichloro-1-2-iodophenoxy
PubChem CID 2776994
IUPAC Name 2,4-dichloro-1-(2-iodophenoxy)benzene
SMILES ClC1=CC(Cl)=C(OC2=CC=CC=C2I)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3OC6H4I
Quantity 1 g
Formula Weight 364.99
Percent Purity 95%
Chemical Name or Material 2, 4-Dichloro-1-(2-iodophenoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.