accessibility menu, dialog, popup

UserName

OOOOH3CH3CCH3CH3MoOO

CAS RN 17524-05-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 328.18
Molecular Formula: C10H16MoO6
InChi Key: FAQSSRBQWPBYQC-UZUXQKAQSA-N
SMILES: O=[Mo]=O.C\C(O)=C\C(C)=O.C\C(O)=C/C(C)=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)