missing translation for 'onlineSavingsMsg'
Learn More

Molybdenyl acetylacetonate, 97%

Product Code. 10656802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656802 50 g Glass bottle 50g
11914831 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656802 Supplier Thermo Scientific Acros Supplier No. 211140500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17524-05-9 | C10H14MoO6 | 326.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17524-05-9
Molecular Formula C10H14MoO6
Molecular Weight (g/mol) 326.17
MDL Number MFCD00011506
InChI Key SKMUJBBRXZPAJY-VGKOASNMSA-L
Synonym 2, 4-Pentanedione, metal derivative
IUPAC Name dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 184.0°C
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 97%
Solubility Information Solubility in water: 85g/L (20°C). Other solubilities: 0.31 g/100g alcohol (20°C), 1.84 g/100g benzene (20°C)
Formula Weight 326.15
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Molybdenyl acetylacetonate
compliance-icons
Product Identifier
  • Molybdenyl acetylacetonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.