missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%

Product Code. 11399628
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399628 10 g 10g
11309638 2 g 2g
11389628 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399628 Supplier Thermo Scientific Alfa Aesar Supplier No. 044140.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17524-05-9 | C10H14MoO6 | 326.17 g/mol

Forms a functional model compound for the study of oxotransferase molybdenum enzymes. Mild catalyst for the deprotection of acetals in high yield. Catalyzes high yield oxidation of secondary alcohols to ketones with sodium percarbonate and phase-transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17524-05-9
Molecular Formula C10H14MoO6
Molecular Weight (g/mol) 326.17
MDL Number MFCD00011506
InChI Key SKMUJBBRXZPAJY-VGKOASNMSA-L
Synonym Bis(acetylacetonato)dioxomolybdenum(VI); Molybdenyl acetylacetonate
IUPAC Name dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼185°C (decomposition)
Color Yellow
Physical Form Powder
Quantity 10 g
Formula Weight 326.16
Percent Purity 99%
Sensitivity Air Sensitive
Chemical Name or Material Molybdenum (VI) oxide bis (2,4-pentanedionate)
compliance-icons
Product Identifier
  • Molybdenum(VI) oxide bis(2,4-pentanedionate)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Molybdenum(VI) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.