accessibility menu, dialog, popup

UserName

BrH3CP

CAS RN 1779-49-3

IUPAC Name: methyltriphenylphosphanium bromide
Synonyms: methyltriphenylphosphanium bromide
Molecular Weight (g/mol): 357.23
Molecular Formula: C19H18BrP
InChi Key: LSEFCHWGJNHZNT-UHFFFAOYSA-M
SMILES: [Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (3)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (7)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)