missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium bromide, 98+%

Product Code. 11492588
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492588 50 g 50g
11402598 250 g 250g
11482588 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492588 Supplier Thermo Scientific Alfa Aesar Supplier No. A15878.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1779-49-3 | C19H18BrP | 357.23 g/mol

Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium bromide is used for methylenation through the Wittig reaction. It is utilized in the synthesis of an enyne and 9-isopropenyl -phenanthrene by using sodium amide as reagent. It is a lipophilic molecule and used to deliver molecules to specific cell components. Further, it acts as an antineoplastic agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1779-49-3
Molecular Formula C19H18BrP
Molecular Weight (g/mol) 357.23
MDL Number MFCD00011804
InChI Key LSEFCHWGJNHZNT-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
PubChem CID 74505
IUPAC Name methyl(triphenyl)phosphanium;bromide
SMILES [Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifikationer

Melting Point 231°C to 234°C
Assay Percent Range ≥98%
Quantity 50 g
UN Number UN2811
Beilstein 3599467
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 357.24
Percent Purity ≥98%
Chemical Name or Material Methyltriphenylphosphonium bromide
compliance-icons compliance-icons
Produktbeteckning
  • Methyl triphenylphosphonium bromide
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Faroangivelser
  • H301-Toxic if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Kompletterande information
  • MIXTURE LIST-Contain: Methyl triphenylphosphonium bromide

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.