accessibility menu, dialog, popup

UserName

OOFCl

CAS RN 177985-32-9

IUPAC Name: 2-(2-chloro-4-fluorophenyl)acetate
Synonyms: 2-(2-chloro-4-fluorophenyl)acetate
Molecular Weight (g/mol): 187.57
Molecular Formula: C8H5ClFO2
InChi Key: CDUSPKFHAVQVRQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=C(F)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results