missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorophenylacetic acid, 97%

Product Code. 11489917
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489917 25 g 25g
11479917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489917 Supplier Thermo Scientific Alfa Aesar Supplier No. B22872.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 177985-32-9 | C8H6ClFO2 | 188.582 g/mol

2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177985-32-9
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00236028
InChI Key CDUSPKFHAVQVRQ-UHFFFAOYSA-N
PubChem CID 2725061
IUPAC Name 2-(2-chloro-4-fluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1F)Cl)CC(=O)O

Specifications

Melting Point 102°C to 105°C
Quantity 25 g
Synonym 2-chloro-4-fluorophenylacetic acid, 2-2-chloro-4-fluorophenyl acetic acid, 2-chloro-4-fluorophenyl acetic acid, 2-2-chloro-4-fluoro-phenyl acetic acid, rarechem al bo 0843, timtec-bb sbb003675, labotest-bb lt00441150, benzeneacetic acid,2-chloro-4-fluoro, 2-chloro-4-fluoro-phenyl-acetic acid, benzeneacetic acid, 2-chloro-4-fluoro
Solubility Information Slightly soluble in water.
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.