accessibility menu, dialog, popup

UserName

CAS RN 1782235-14-6

CH3NHNNNClClHNNHNNHClHClHCl

CAS RN 1782235-14-6

IUPAC Name: 6-[(2-{[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile trihydrochloride
Synonyms: 6-[(2-{[4-(2,4-dichlorophenyl)-5-(4-methyl-3H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile trihydrochloride
Molecular Weight (g/mol): 574.72
Molecular Formula: C22H21Cl5N8
InChi Key: DSFVSCNMMZRCIA-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.CC1=CN=C(N1)C1=CN=C(NCCNC2=NC=C(C=C2)C#N)N=C1C1=C(Cl)C=C(Cl)C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)