Learn More
CHIR 99021 trihydrochloride, Tocris Bioscience™
GREENER_CHOICE
Hydrochloride salt of CHIR 99021 (Cat. No. 4423); selective GSK-3 inhibitor
Brand:  Tocris Bioscience 4953/50
Encompass
Product Code. 15243327
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Hydrochloride salt of CHIR 99021 (Cat. No. 4423), a highly selective GSK-3 inhibitor.
Specifications
Specifications
1782235-14-6
50mg
DSFVSCNMMZRCIA-UHFFFAOYSA-N
78243722
>99%
C22H21Cl5N8
chir-99021 trihydrochloride, chir 99021 trihydrochloride, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino-3-pyridinecarbonitrile trihydrochloride
574.72
574.72
CHIR 99021 trihydrochloride